Läs mer
Icomethasone, TRC
Produktkod. 30403669
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30403669 10 mg
30393424 500 mg
30402479 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30403669 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI16410010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4647-20-5
Molekylformel C22 H29 Cl O5
Molekylvikt (g/mol) 408.92
Synonym Pregna-1,4-diene-3,20-dione, 9-chloro-11,17,21-trihydroxy-16-methyl-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 9-chloro-11β,17,21-trihydroxy-16α-methyl- (7CI,8CI), (11β,16α)-9-Chloro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, Icometasone, Icomethasone, Mometasone Furoate Imp. K (EP)
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material Icomethasone
Kvantitet 10 mg
InChI formel InChI=1S/C22H29ClO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1
Formel vikt 408.1704