Learn More
ID2 Monoclonal Antibody (ILCID2), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1480.00 SEK - 4030.00 SEK
Specifications
Antigen | ID2 |
---|---|
Clone | ILCID2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933254
|
Affymetrix eBioscience
25-9475-80 |
25 μg |
1480.00 SEK
25µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943254
|
Affymetrix eBioscience
25-9475-82 |
100 μg |
4030.00 SEK
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This ILCID2 monoclonal antibody recognizes mouse and human ID2. This ILCID2 antibody requires Foxp3/Transcription Factor Staining Buffer set and will perform poorly when used with standard IC Fixation & Permeabilization Buffer Set and protocol. Applications Reported: This ILCID2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ILCID2 antibody has been tested by flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product No. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No.
ID2 Monoclonal antibody specifically detects ID2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
ID2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Ac2-300; AI255428; bHLHb26; C78922; cell growth-inhibiting gene 8; class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID2; DNA-binding protein inhibitor ID-2; GIG8; helix-loop-helix protein ID2; Id2; Id-2; ID2A; ID2H; Idb2; Inhibitor of differentiation 2; inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein; inhibitor of DNA binding 2, HLH protein; MGC26389; OTTHUMP00000140258; OTTHUMP00000200297 | |
ID2 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
ILCID2 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human, Mouse | |
P41136, Q02363 | |
15902, 3398 | |
Purified recombinant fragment of human ID2, corresponding to amino acids 1-134 expressed in E. coli | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ID2 |