missing translation for 'onlineSavingsMsg'
Learn More

IDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12332-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16839993

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This nuclear enzyme is involved in processes such as chromosome condensation, chromatid separation, and the relief of torsional stress that occurs during DNA transcription and replication. It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another, thus altering the topology of DNA. Two forms of this enzyme exist as likely products of a gene duplication event. The gene encoding this form, beta, is localized to chromosome 3 and the alpha form is localized to chromosome 17. The gene encoding this enzyme functions as the target for several anticancer agents and a variety of mutations in this gene have been associated with the development of drug resistance. Reduced activity of this enzyme may also play a role in ataxia-telangiectasia. Alternative splicing of this gene results in two transcript variants; however, the second variant has not yet been fully described.
Specifications

Specifications

IDH1
Polyclonal
Unconjugated
IDH1
IDCD, IDH, IDH1, IDP, IDPC, NADP(+) specific ICDH, Oxalosuccinate decarboxylase, PICD
Rabbit
Antigen Affinity Chromatography
RUO
15926, 24479, 3417
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75874, O88844, P41562
IDH1
IDH1 Fusion Protein Ag2986
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers