missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23254-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to IDH2.
Plays a role in intermediary metabolism and energy production. It may tightly associate or interact with the pyruvate dehydrogenase complex.Specifications
IDH2 | |
Polyclonal | |
Unconjugated | |
IDH2 | |
ICD M, IDH, IDH2, IDHM, IDP, IDPM, mNADP IDH, NADP(+) specific ICDH, Oxalosuccinate decarboxylase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269951, 3418, 361596 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48735, P54071, P56574 | |
Idh2 | |
IDH2 Fusion Protein Ag19748 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |