missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH3G Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25848-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predomitly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the gamma subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. This gene is a candidate gene for periventricular heterotopia. Several alternatively spliced transcript variants of this gene have been described, but only some of their full length natures have been determined.Specifications
IDH3G | |
Polyclonal | |
Unconjugated | |
IDH3G | |
H IDHG, IDH3G | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15929, 3421 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51553, P70404 | |
IDH3G | |
IDH3G Fusion Protein Ag23113 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |