missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IDI1 (Human) Recombinant Protein

Human IDI1 full-length ORF ( AAH05247.1, 1 a.a. - 228 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00003422-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16129804

  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MMPEINTNHLDKQQVQLLAEMCILIDENDNKIGAETKKNCHLNENIEKGLLHRAFSVFLFNTENKLLLQQRSDAKITFPGCFTNTCCSHPLSNPAELEESDALGVRRAAQRRLKAELGIPLEEVPPEEINYLTRIHYKAQSDGIWGEHEIDYILLVRKNVTLNPDPNEIKSYCYVSKEELKELLKKAASGEIKITPWFKIIAATFLFKWWDNLNHLNQFVDHEKIYRM
Specifications

Specifications

AAH05247.1
isopentenyl-diphosphate delta isomerase 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IDI1
GST
3422
Wheat germ expression system
10 μg
IPP1, IPPI1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers