Learn More
IDO Monoclonal Antibody (eyedio), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
3325.00 SEK
Specifications
Antigen | IDO |
---|---|
Clone | eyedio |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15538666
|
Affymetrix eBioscience
25-9477-42 |
100 Tests |
3325.00 SEK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This eyedio monoclonal antibody reacts with human indoleamine-2,3-dioxygenase (IDO, INDO, IDO1), an intracellular enzyme that catalyzes the degradation of tryptophan to kynurenines. IDO is expressed in a wide variety of tissues and cells, including macrophages, plasmacytoid dendritic cells, and several cell lines. IDO can be induced in many different cell types by IFN gamma or other inflammatory stimuli. Expression of IDO in antigen presenting cells and tumors is thought to mediate immune suppression through depletion of this essential amino acid and/or by the creation of tryptophan metabolites that cause apoptosis of T cells and induction of regulatory T cells. Applications Reported: This eyedio antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eyedio antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This antibody can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
IDO Monoclonal antibody specifically detects IDO in Human samples. It is validated for Flow Cytometry.Specifications
IDO | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P14902 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
eyedio | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
3620 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.