Läs mer
Invitrogen™ IFI30 (Precursor) Recombinant Rabbit Monoclonal Antibody (3H6L14)
Produktkod. 17106298
Change view
Kvantitet:
100 μg
Produktkod. Kvantitet
17106298 100 μg
1 options
Produktkod. 17106298 Leverantör Invitrogen™ Leverantörsnummer 703502
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Rabbit Recombinant Monoclonal Antibody
This antibody is predicted to react with Mouse, Rat.
The protein encoded by this gene is a lysosomal thiol reductase that at low pH can reduce protein disulfide bonds. The enzyme is expressed constitutively in antigen-presenting cells and induced by gamma-interferon in other cell types. This enzyme has an important role in MHC class II-restricted antigen processing.
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen IFI30 (Precursor)
Användningsområden Western Blot, Immunocytochemistry
Klassificering Recombinant Monoclonal
Klona 3H6L14
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.09% sodium azide; pH 7.4
Gen IFI30
Genaccessionsnr. P13284
Gene Alias Gamma-interferon-inducible lysosomal thiol reductase; gamma-interferon-inducible protein IP-30; GILT; IFI30; IFI-30; IFI30, lysosomal thiol reductase; interferon gamma inducible protein 30; interferon gamma-inducible protein 30 preproprotein; interferon, gamma-inducible protein 30; IP30; IP-30; Legumaturain
Gensymboler IFI30
Värd art Rabbit
Immunogen Peptide corresponding to human IF130 [1) aa38-aa56 2)aa233-aa250].
Reningsmetod Protein A
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 10437
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG