missing translation for 'onlineSavingsMsg'
Learn More

IFIH1 Mouse anti-Human, Clone: 3D8B11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66770-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828955

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Melanoma differentiation associated gene 5, MDA5, is a pattern recognition receptor to transmit the signal.Once activated, both proteins signal through IPS-1 activating transcription factors NF- kappaB and IRF-3 and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus, paromyxoviruses, Japanese encephalitis virus. MDA5 is essential for signaling by picornavirus. Both RIG-I and MDA5 are responsible for signaling by West Nile virus.
Specifications

Specifications

IFIH1
3D8B11
Unconjugated
IFIH1
Liquid
IFIH1
IFIH1 Fusion Protein Ag17448
Protein G
RUO
Antibody
Monoclonal
Human
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BYX4
Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, IFIH1, MDA 5, MDA5, RH116
Mouse
IgG1
20 μL
-20°C
Primary
64135
Videos
SDS
Documents
Certificates
Special Offers

Special Offers