Learn More
IFIT3 Mouse anti-Human, Unconjugated, Clone: 2C8B10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67208-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839725
  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The IFIT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IFIT3. This antibody recognizes human antigen. The IFIT3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
IFIT3
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
2C8B10
Unconjugated
150 μL
3437
IgG2a