missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM2/3 Mouse anti-Human, Clone: 2E8D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66081-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody recognizes both IFITM2 and IFITM3.
IFN-induced antiviral protein that mediates cellular innate immunity to at least three major human pathogens, namely influenza A H1N1 virus, West Nile virus (WNV), and dengue virus (WNV), by inhibiting the early step(s) of replication.Specifications
IFITM2/3 | |
Monoclonal | |
0.85 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01628, Q01629 | |
IFITM2, IFITM3 | |
IFITM2/3 Fusion Protein Ag17863 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2E8D12 | |
Unconjugated | |
IFITM3 | |
1 8U, IFITM3, IP15 | |
Mouse | |
Protein G | |
RUO | |
10410, 10581 | |
-20°C | |
Liquid |