Learn More
IFN gamma Monoclonal Antibody (B308), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience BMS1019
Additional Details : Weight : 0.09500kg
Description
Description: IFN-gamma, also called Type II interferon, is a homodimeric glycoprotein containing approximately 21 to 24 kD subunits. The human IFN-gamma gene, situated on chromosome 12, contains three introns; the four exons code for a polypeptide of 166 amino acids, 20 of which constitute the signal peptide. In contrast to IFN-aalpha and IFN-beta synthesis, which can occur in any cell, production of IFN-gamma is a function of T cells and NK cells. IFN-gamma has antiviral and antiparasitic activities and also inhibits the proliferation of a number of normal and transformed cells. IFN-gamma synergises with TNF-alpha and TNF-beta in inhibiting the proliferation of various cell types. Applications Tested: ELISA, Immunoprecipitation.
IFN gamma Monoclonal antibody specifically detects IFN gamma in Human samples. It is validated for ELISA, Immunoprecipitation.Specifications
IFN gamma | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
P01579 | |
IFNG | |
Rec. Human IFN gamma | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation | |
B308 | |
Unconjugated | |
IFNG | |
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g | |
Mouse | |
Affinity chromatography | |
RUO | |
3458 | |
-20°C | |
Liquid |
For Research Use Only.