missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ IFN gamma Monoclonal Antibody (B27), PE

Produktkod. 10517983
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 μL
3 mL
Förpackningsstorlek:
Varje
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
10517983 3 mL Varje
10507983 500 μL Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy

For Research Use Only. All usage must comply with product instructions.

Produktkod. 10517983 Leverantör Invitrogen™ Leverantörsnummer MHCIFG043
  • ONLINE EXCLUSIVE PRICE
    10280.00 SEK
    8738.00 SEK / 3ml
    Save 1542.00 SEK
    15% Av
 Begränsad artikel
Beräknad leverans: 13-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For Research Use Only. All usage must comply with product instructions.

Mouse Monoclonal Antibody

IFN gamma Monoclonal Antibody for Flow

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen IFN gamma
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona B27
Koncentration 0.1 mg/mL
Konjugera PE
Formulering PBS with sucrose, 4mg/mL BSA and 0.1% sodium azide
Gen IFNG
Genaccessionsnr. P01579
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g
Gensymboler IFNG
Värd art Mouse
Immunogen Human IFN-gamma.
Reningsmetod Purified
Kvantitet 3 mL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 3458
Målarter Human
Innehåll och lagring 4°C
Produkttyp Antibody
Form Liquid
Isotyp IgG1 κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.