missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ IFNLR1 Monoclonal Antibody (601106)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524271
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
In direct ELISAs, no cross-reactivity with recombinant mouse IL-28 alpha is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
The protein encoded by this gene belongs to the class II cytokine receptor family. This protein forms a receptor complex with interleukine 10 receptor, beta (IL10RB). The receptor complex has been shown to interact with three closely related cytokines, including interleukin 28A (IL28A), interleukin 28B (IL28B), and interleukin 29 (IL29). The expression of all three cytokines can be induced by viral infection. The cells overexpressing this protein have been found to have enhanced responses to IL28A and IL29, but decreased response to IL28B. Three alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifikationer
| IFNLR1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8IU57 | |
| Ifnlr1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-28 R alpha/IFN-lambda R1 Arg21-Ala228. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 601106 | |
| Unconjugated | |
| Ifnlr1 | |
| class II cytokine receptor 12; class II cytokine receptor CRF2/12; CRF2/12; CRF2-12; Cytokine receptor class-II member 12; cytokine receptor family 2 member 12; IFN-lambda R1; IFN-lambda receptor 1; IFN-lambda-R1; IFNLR; IFNLR1; IL-28 receptor subunit alpha; IL-28R1; Il28ra; IL-28RA; IL-28R-alpha; Interferon lambda receptor 1; interferon lambda, receptor 1; interferon, lambda receptor 1; interferon-lambda receptor 1; interleukin 28 alpha receptor; interleukin 28 receptor A; interleukin 28 receptor alpha; interleukin 28 receptor, alpha (interferon, lambda receptor); interleukin or cytokine receptor 2; Interleukin-28; interleukin-28 receptor subunit alpha; LICR2; Likely interleukin or cytokine receptor 2; RGD1562689 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 163702 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering