missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT43 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24338-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
IFT43, encoded by C14ORF179, is a subunit of the intraflagellar transport complex A (IFT-A) of primary cilia. The IFT-A is implicated in retrograde ciliary transport along axonemal microtubules. Defects in subunits of IFT-A has been associated with skeletal ciliopathies, including Sensenbrenner syndrome, Jeune syndrome, and Ellis-van-Creveld syndrome. Mutations in C14ORF179 has been linked to Sensenbrenner syndrome. (21378380).Specifications
IFT43 | |
Polyclonal | |
Unconjugated | |
IFT43 | |
C14orf179, IFT43 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112752 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96FT9 | |
IFT43 | |
IFT43 Fusion Protein Ag19434 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |