missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT43 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | IFT43 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882815
|
Proteintech
24338-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16872815
|
Proteintech
24338-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
IFT43, encoded by C14ORF179, is a subunit of the intraflagellar transport complex A (IFT-A) of primary cilia. The IFT-A is implicated in retrograde ciliary transport along axonemal microtubules. Defects in subunits of IFT-A has been associated with skeletal ciliopathies, including Sensenbrenner syndrome, Jeune syndrome, and Ellis-van-Creveld syndrome. Mutations in C14ORF179 has been linked to Sensenbrenner syndrome. (21378380).Specifications
IFT43 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q96FT9 | |
112752 | |
IFT43 Fusion Protein Ag19434 | |
Primary | |
-20°C | |
IFT43 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C14orf179, IFT43 | |
IFT43 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |