missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT88 Mouse anti-Human, Mouse, Rat, Clone: 4A4G5, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | IFT88 |
---|---|
Clone | 4A4G5 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887245
|
Proteintech
60227-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877245
|
Proteintech
60227-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody was raised against the C-terminal region of human IFT88 and can detect the endogenous level of IFT88.
This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
IFT88 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
IFT88, TPR repeat protein 10, TTC10 | |
IFT88 | |
IgG1 | |
Protein G | |
Antibody |
4A4G5 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q13099, Q61371 | |
21821, 305918, 8100 | |
IFT88 Fusion Protein Ag4980 | |
Primary | |
-20°C | |
IFT88 |