missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ IgE (ε -Heavy Chain), Rabbit, Ready to use, Polyclonal, Epredia™

IgE (ε-Heavy Chain), Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia™ RB-1435-R7

Additional Details : Weight : 0.01200kg

Product Code. 12612907

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Research Areas: Immunoglobulins; The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Clone: Polyclonal

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: IgE isolated from a pool of normal human sera

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

IgE (epsilon-Heavy Chain)
Polyclonal
Prediluted antibody which is ready-to-use for staining of formalin-fixed, paraffin-embedded tissues.
IgE isolated from a pool of normal human sera
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
7 mL
Immunoglobulins
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers