missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
IgE Monoclonal Antibody (Ige21), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2075.00 SEK - 4420.00 SEK
Specifikationer
| Antigen | IgE |
|---|---|
| Klona | Ige21 |
| Koncentration | 0.2 μg/Test |
| Användningsområden | Flow Cytometry |
| Klassificering | Monoclonal |
Beskrivning
IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.Specifikationer
| IgE | |
| 0.2 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide, pH 11.2 | |
| HCAK1, IgA1, IgG3, IGHA1, IGHA2, IGHG1, IGHG2, IGHG3, IGHG4, IGKC, IGKCD, IGLC, IGLC1, Immunoglobulin E, immunoglobulin heavy constant alpha 1, immunoglobulin heavy constant alpha 2 (A2m marker), immunoglobulin heavy constant gamma 1 (G1m marker), immunoglobulin heavy constant gamma 2 (G2m marker), immunoglobulin heavy constant gamma 3 (G3m marker), immunoglobulin heavy constant gamma 4 (G4m marker), immunoglobulin kappa constant, immunoglobulin lambda constant 1, ImmunoglobulinE, Km | |
| IGHE | |
| Primary | |
| 4°, C, store in dark, DO NOT FREEZE! |
| Ige21 | |
| Flow Cytometry | |
| NovaFluor Blue 510 | |
| Mouse | |
| Human | |
| P01854 | |
| 3497 | |
| IgG1 κ | |
| Affinity chromatography | |
| IGHE |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel