missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
IGF1R alpha Monoclonal Antibody (24-60), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513817
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA5-13817 targets IGF-1 Receptor alpha in IA, ICC/IF, WB and IP applications and shows reactivity with Human and Mouse samples. The MA5-13817 immunogen is iGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA.
IGF1R alpha Monoclonal antibody specifically detects IGF1R alpha in Human, Mouse samples. It is validated for Immunocytochemistry, Immunoprecipitation, Inhibition Assays, Western Blot.
Specifikationer
| IGF1R alpha | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P08069, Q60751 | |
| Igf1r | |
| IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a κ |
| Immunocytochemistry, Immunoprecipitation, Inhibition Assays, Western Blot | |
| 24-60 | |
| Unconjugated | |
| Igf1r | |
| A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 16001, 3480 | |
| 4° C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering