missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGF1R Recombinant Human Protein
IGF1R is also known as Insulin-like Growth Factor 1 (IGF-1) Receptor.
Brand: Invitrogen™ PR4654A
Additional Details : Weight : 0.01000kg
Description
IGF1R is a receptor tyrosine kinase activated by IGF-1 and IGF-2. IGFR has been implicated in a variety of cancers.
Specifications
NP_000866 | |
50mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
IGF-1R | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
48.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
Insulin & IGF Receptor Kinase Family | |
Recombinant | |
Baculovirus |