missing translation for 'onlineSavingsMsg'
Learn More

Armenian Hamster IgG, eFluor 660, clone: eBio299Arm, Isotype Control, eBioscience™

Armenian Hamster Isotype Control

644.00 SEK - 1390.00 SEK

Specifications

Antigen Armenian Hamster IgG
Clone eBio299Arm
Target Species Not Applicable
Conjugate eFluor 660
Isotype IgG
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15559966
View Documents
Affymetrix eBioscience
50-4888-80
25μg
644.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment
02-03-2021
Add to basket
 
15569966
View Documents
Affymetrix eBioscience
50-4888-82
100μg
1390.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment
02-03-2021
Add to basket
 
Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use isotype control at the same concentration as experimental antibody.eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Armenian Hamster IgG
Not Applicable
IgG
Monoclonal
Control
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
RUO
eBio299Arm
eFluor 660
Control, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Liquid
Isotype Control
Store at 2-8°C. Do not freeze. Light-sensitive material.
SDS

For Research Use Only.