Learn More
Rabbit IgG Isotype Control, FITC, eBioscience™, Invitrogen™
Rabbit Isotype Control
Brand: Affymetrix eBioscience 11-4614-80
590.69 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.25000kg
Description
Description: The rabbit IgG polyclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: The Rabbit IgG isotype control has been reported for use as a negative control for immunofluorescent staining and flow cytometric analysis. Applications Tested: This isotype control antibody has been tested by flow cytometric analysis. Please use at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.Specifications
Rabbit IgG | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rabbit | |
25 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |
Control, Flow Cytometry | |
FITC | |
Immunoglobulin G; ImmunoglobulinG; Not Applicable | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
For Research Use Only.