Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 41-4714-82
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control immunoglobulin. Applications Reported: This P3.6.2.8.1 antibody has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: This Mouse IgG1 K Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized MDCK cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human brain tissue. Use isotype control at the same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Mouse IgG1 kappa | |
P3.6.2.8.1 | |
eFluor 570 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
For Research Use Only.