Learn More
Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Secondary Antibody
976.00 SEK - 2110.00 SEK
Specifications
Antigen | Rat IgG2a |
---|---|
Clone | r2a-21B2 |
Concentration | 0.2 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15548946
|
Affymetrix eBioscience
41-4817-80 |
25 μg |
976.00 SEK
25µg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15558946
|
Affymetrix eBioscience
41-4817-82 |
100 μg |
2110.00 SEK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody r2a-21B2 recognizes Rat IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2a or any mouse isotype antibodies. Applications Reported: This R2a-21B2 antibody has been reported for use in immunocytochemistry, and immunohistochemical staining of frozen and formalin-fixed paraffin embedded tissue sections. Applications Tested: This R2a-21B2 antibody has been tested by immunocytochemistry on fixed and permeabilized cells that have been stained first with a rat IgG2a anti-mouse antibody. This can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Rat IgG2a Monoclonal antibody specifically detects Rat IgG2a in Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Rat IgG2a | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
r2a-21B2 | |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
eFluor 570 | |
Mouse | |
Rat | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.