Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), APC, eBioscience™, Invitrogen™
Mouse Isotype Control
994.00 SEK - 1676.00 SEK
Specifications
Antigen | Mouse IgG2b kappa |
---|---|
Clone | eBMG2b |
Applications | Control, Flow Cytometry |
Conjugate | APC |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15330730
|
Affymetrix eBioscience
17-4732-81 |
50 μg |
1214.00 SEK
50µg |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
15558096
|
Affymetrix eBioscience
17-4732-41 |
25 Tests |
994.00 SEK
25 tests |
In Stock
Log in to see stock. |
|||||
15568096
|
Affymetrix eBioscience
17-4732-42 |
100 Tests |
1676.00 SEK
100 tests |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: APC Mouse IgG2b Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: APC Mouse IgG2b Isotype Control is offered in 2 formats: - μg size: has been tested by flow cytometric analysis of normal human peripheral blood cells. This should be used at the same concentration as the experimental antibody. - test size: has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at test size: 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG2b kappa | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBMG2b | |
APC | |
Mouse | |
Not Applicable | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.