Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Isotype Control
1258.00 SEK - 3300.00 SEK
Specifications
Antigen | Mouse IgG2b kappa |
---|---|
Clone | eBMG2b |
Conjugate | eFluor 570 |
Form | Liquid |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15578936
|
Affymetrix eBioscience
41-4732-82 |
100 μg |
3300.00 SEK
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15568936
|
Affymetrix eBioscience
41-4732-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: This eBMG2b antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, immunohistochemical staining, and immunocytochemistry. Applications Tested: This eBMG2b antibody has been tested by immunocytochemistry on fixed and permeabilized cells. Use isotype control at same concentration as experimental antibody. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Mouse IgG2b kappa | |
eFluor 570 | |
Mouse | |
IgG2b κ | |
Affinity chromatography |
eBMG2b | |
Liquid | |
RUO | |
Isotype Control (Primary) |
For Research Use Only.