Learn More
Mouse IgG3 Isotype Control (B10), PE, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 12-4742-41
Additional Details : Weight : 0.23750kg
Description
Description: The mouse IgG3 lambda is useful as an isotype control immunoglobulin. Applications Reported: This mouse IgG3 isotype control has been reported for use in flow cytometric analysis. Applications Tested: This mouse IgG3 isotype control has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 10 μL per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. . Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Description: The mouse IgG3 lambda is useful as an isotype control immunoglobulin.Applications Reported: This mouse IgG3 isotype control has been reported for use in flow cytometric analysis.Applications Tested: This mouse IgG3 isotype control has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 10 μL per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. .Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Mouse IgG3 | |
B10 | |
PE | |
IgG3 | |
Affinity chromatography | |
RUO | |
None | |
Liquid |
Flow Cytometry | |
10μl/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG3 λ |
For Research Use Only.