missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGG3 Rabbit anti-Mouse, Clone: RM218, Abnova™
Rabbit monoclonal antibody raised against native mouse IgG3.
Brand: Abnova MAB12770.100ug
Additional Details : Weight : 0.00010kg
Specifications
IGG3 | |
Monoclonal | |
Unconjugated | |
ELISA (0.005 - 0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5 - 2 ug/mL) Immunohistochemistry (0.5 - 2 ug/mL) Immunoprecipitation Western Blot (0.1 - 0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
IGHG3/IgG3/MGC68300 | |
Rabbit | |
Protein A affinity purification | |
RUO | |
380795 | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RM218 | |
Rabbit monoclonal antibody raised against native mouse IgG3. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
AI324046 | |
Native purified IgG3 from mouse hybridoma. | |
100 μg | |
Primary | |
This antibody reacts to the Fab region of mouse IgG3. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgM, IgA, IgE, human IgG, or rat IgG. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |