missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGHM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | IGHM |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887793
|
Proteintech
11016-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877793
|
Proteintech
11016-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody detects the heavy chain of human IgM (∽75 kDa).
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.Specifications
IGHM | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P01871 | |
3507 | |
IGHM Fusion Protein Ag1459 | |
Primary | |
-20°C | |
IGHM |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IGHM, MU, VH | |
IGHM | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |