Learn More
IGHM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11016-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody detects the heavy chain of human IgM (∽75 kDa).
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.Specifications
IGHM | |
Polyclonal | |
Unconjugated | |
IGHM | |
IGHM, MU, VH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3507 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01871 | |
IGHM | |
IGHM Fusion Protein Ag1459 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |