Learn More
IGSF9B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552742
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LEAPLSSGKV SPESIRTLRA PSESSDDQGQ PAAKRMLSPT REKELSLYKK TKRAISSKKY SVAKAEAEAE ATTPIELISR GPDGRFVMDP AEMEPSLKSR RIEGFPFAEE TDMYPEFRQS DEENEDPLVP TSVAALKSQL TPLSSSQE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
IGSF9B Polyclonal antibody specifically detects IGSF9B in Human samples. It is validated for Immunohistochemistry.Specifications
IGSF9B | |
Polyclonal | |
Unconjugated | |
IGSF9B | |
Gm508; IgSF9B; igsf9l; igsf9l protein; im:7148508; immunoglobulin superfamily member 9B; immunoglobulin superfamily, member 9b; immunoglobulin superfamily, member 9b; protein turtle homolog A; Kiaa1030; LOW QUALITY PROTEIN: protein turtle homolog B; mKIAA1030; Protein turtle homolog B; RGD1564717; si:dkey-49h9.6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22997 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPX0 | |
IGSF9B | |
Recombinant protein corresponding to Human IGSF9B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only