missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKBKB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20979-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to IKBKB.
Inhibitor of nuclear factor kappa-B kinase subunit beta (IKK beta) is a serine kinase that plays an essential role in the NF-kaapa-B signaling pathway which is activated by mulitple stimuli such as inflammatory cytokines, bacterial, or viral products, DNA damages, or other cellular stresses. IKK beta acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Mutations affecting the IKK beta gene can result in Immunodeficiency 15 (IMD15).Specifications
IKBKB | |
Polyclonal | |
Unconjugated | |
IKBKB | |
I kappa B kinase 2, I kappa B kinase beta, IKBKB, IKK B, IKK beta, IKK2, IKKB, IkappaB, NFKBIKB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16150, 3551 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14920, O88351 | |
IKBKB | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |