missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IKBKE (IKK Epsilon), GST Tag Recombinant Protein
Human IKBKE (IKK Epsilon), GST Tag Recombinant Protein
5380.00 SEK - 128700.00 SEK
Specifications
Accession Number | NP_054721 |
---|---|
Gene Alias | IKBKE (IKK Epsilon) |
KinaseFamily | IKK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10639633
|
Invitrogen™
PV4875 |
10μg |
5380.00 SEK
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448643
|
Invitrogen™
PR8031B |
100μg |
20240.00 SEK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498763
|
Invitrogen™
PV4876 |
1mg |
128700.00 SEK
1mg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Over expression of IKBKE has been observed in breast cancer cells, and suppression of IKBKE induces cell death.
Specifications
NP_054721 | |
IKK Kinase Family | |
Recombinant | |
Baculovirus |
IKBKE (IKK Epsilon) | |
Human | |
GST Tag |