missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
IKK alpha Antibody (14A231), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27409PE
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IKK alpha Monoclonal specifically detects IKK alpha in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifikationer
| IKK alpha | |
| Monoclonal | |
| PE | |
| O15111 | |
| CHUK | |
| This antibody was raised against a His-tagged full-length human IKK alpha protein. | |
| 0.1 mL | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase, Signal Transduction | |
| 1147 | |
| Human, Mouse, Rat, Primate | |
| Purified |
| Flow Cytometry | |
| 14A231 | |
| Flow Cytometry, Flow (Intracellular) | |
| conserved helix-loop-helix ubiquitous kinaseIKBKA, EC 2.7.11, EC 2.7.11.10, IkappaB kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, IkBKA, IKK1I-kappa-B kinase-alpha, IKK-A, IKKAIkB kinase alpha subunit, IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha, NFKBIKAIKK-a kinase, Nuclear factor NFkappaB inhibitor kinase alpha, Nuclear factor NF-kappa-B inhibitor kinase alpha, TCF16, TCF-16, Transcription factor 16 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| An 85 kDa band should be observed. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion