missing translation for 'onlineSavingsMsg'
Learn More

IKZF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13554-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881824

  • 1656.73 SEK / 20µL
Estimated Shipment: 26-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predomitly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifications

Specifications

IKZF2
Polyclonal
Unconjugated
IKZF2
HELIOS, IKZF2, Zinc finger protein Helios, ZNF1A2, ZNFN1A2
Rabbit
Antigen Affinity Chromatography
RUO
22779, 22807, 301476
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P81183, Q9UKS7
Ikzf2
IKZF2 Fusion Protein Ag4357
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers