missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKZF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19055-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
IKZF3 is a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.Specifications
IKZF3 | |
Polyclonal | |
Unconjugated | |
IKZF3 | |
AIO, AIOLOS, IKZF3, Zinc finger protein Aiolos, ZNFN1A3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22780, 22806, 303511 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08900, Q9UKT9 | |
Ikzf3 | |
IKZF3 Fusion Protein Ag5537 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |