missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ IL-13 Monoclonal Antibody (eBio13A), eFluor™ 660, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

1630.00 SEK - 3840.00 SEK

Specifikationer

Antigen IL-13
Klona eBio13A
Koncentration 0.2 mg/mL
Användningsområden Flow Cytometry
Klassificering Monoclonal
Se fler specifikationer

Produkter 2
Produktkod Brand Kvantitet Pris Kvantitet och tillgänglighet  
Produktkod Brand Kvantitet Pris Kvantitet och tillgänglighet  
15560017
Visa dokument
Invitrogen™
50-7133-80
25 μg
1630.00 SEK
25µg
Se alternativa produkter
Spara upp till 
»
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
15570017
GREENER_CHOICE
Visa dokument
Invitrogen™
50-7133-82
100 μg
3840.00 SEK
100µg
Se alternativa produkter
Spara upp till 
»
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning

Beskrivning

Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12) Applications Reported: This eBio13A antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBio13A antibody has been tested by intracellular staining and flow cytometric analysis of Th2 polarized mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Specifikationer

Specifikationer

IL-13
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Il13
Primary
4°C, store in dark, DO NOT FREEZE!
Il13
eBio13A
Flow Cytometry
eFluor 660
Rat
Mouse
P20109
16163
IgG1 κ
Affinity chromatography
Antibody
Filmer
SDS
Dokument

Dokument

Produktspecifikationer

For Research Use Only.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.