missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13 Rat anti-Mouse, eFluor® 660, Clone: eBio13A, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 50-7133-82
Additional Details : Weight : 0.09500kg
Description
Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
IL-13 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P20109 | |
Interleukin-13, ALRH antibody, NC30 antibody, P600 antibody | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Mouse |
Flow Cytometry | |
eBio13A | |
eFluor™ 660 | |
Il13 | |
Liquid | |
Il13 | |
IgG1, kappa | |
100 μg | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
16163 |
For Research Use Only.