Läs mer
Invitrogen™ IL-15 Monoclonal Antibody (AIO.3), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
24040.00 SEK - 158800.00 SEK
Specifikationer
| Antigen | IL-15 |
|---|---|
| Klona | AIO.3 |
| Koncentration | 1 mg/mL |
| Användningsområden | Functional Assay, Neutralization |
| Klassificering | Monoclonal |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
15507816
|
Invitrogen™
16-7154-38 |
5 mg |
24040.00 SEK
5mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
17151492
|
Invitrogen™
16-7154-025 |
25 mg |
105300.00 SEK
25mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
17161492
|
Affymetrix eBioscience
16-7154-050 |
50 mg | |||||||
Beskrivning
The AIO.3 antibody reacts with mouse IL-15. This is a neutralizing antibody. Mouse IL-15 is a 14-15 kDa factor, previously known as IL-T. IL-15 is produced by a variety of cells and has a role in the T cell-mediated immune response and the central nervous system. IL-15 binds to the beta and gamma subunits of the IL-2 receptor; however, these two cytokines have distinct biological activities. IL-15 is a specific maturation factor for NK cells and mimics the BM microenvironment for NK cell development in vitro. It is reported that IL-15 delivers an anti-apoptotic and survival signal for NK, T, and B cells.
IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.Specifikationer
| IL-15 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 16168 | |
| Primary | |
| 4°C | |
| IL15 |
| AIO.3 | |
| Functional Assay, Neutralization | |
| Functional Grade | |
| Rat | |
| Mouse | |
| P48346 | |
| IgG2a λ | |
| Affinity Chromatography | |
| Antibody |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.