Läs mer
Invitrogen™ IL-17AF Monoclonal Antibody (20LJS09), PE, eBioscience™

Mouse Monoclonal Antibody
4390.00 SEK
Specifikationer
| Antigen | IL-17AF |
|---|---|
| Klona | 20LJS09 |
| Koncentration | 5 μL/Test |
| Innehåll och lagring | 4°C, store in dark, DO NOT FREEZE! |
| Användningsområden | Flow Cytometry |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
15557186
![]() |
Invitrogen™
12-9179-42 |
100 Tests |
4390.00 SEK
100 tester |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
Description: The 20LJS09 monoclonal antibody reacts with human interleukin (IL)-17AF heterodimer. This antibody specifically reacts with this heterodimer and has no reactivity for IL-17A or IL-17F homodimers. IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis. Applications Reported: This 20LJS09 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 20LJS09 antibody has been pre-titrated and tested on 10 day in vitro-polarized Th17 cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.Specifikationer
| IL-17AF | |
| 5 μL/Test | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q16552, Q96PD4 | |
| 112744, 3605 | |
| Primary | |
| Affinity chromatography |
| 20LJS09 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| Il17a | |
| C87042; CANDF6; Ctla8; CTLA-8; cytokine ML-1; Cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; IL17; IL-17; Il17a; IL-17A; IL17F; IL-17F; ILN; Interleukin; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17A; interleukin 17F; interleukin-17A; Interleukin-17F; ML1; ML-1 | |
| Il17a, Il17f | |
| Antibody |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.