Learn More
IL-20RB, clone: 20RNTC, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-1206-80
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15277187
  • 924.00 SEK / 25µg
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
This 20RNTC monoclonal antibody recognizes mouse and human IL-20R2 (interleukin-20 receptor 2, IL-20Rb) in either of its two heterodimeric complexes formed by pairing with IL-20R1 or IL-22R1. IL-20R2 belongs to the class II cytokine receptor family that shares a characteristic pattern of cysteine, proline and tryptophan residues in the extracellular domain. IL-20R2 paired with IL-20R1 forms a receptor that binds IL-20, IL-19, or IL-24. IL-20R2 paired with IL22R1 forms a receptor that binds IL-20 or IL-24. The IL-20R complex signals primarily via the JAK-STAT pathway. IL-20R2 is found predominantly on cells of epithelial origin such as gut and skin and can be upregulated during inflammation. Immunofluorescent staining of A375 cells reveals a punctate staining pattern at the cell membrane, as well as diffuse, cytoplasmic staining that is likely associated with protein synthesis and transport in the endoplasmic reticulum and golgi.
Specifications
Specifications
IL-20 Receptor 2
20RNTC
Unconjugated
Purified
Rat
25μg
Store at 2-8°C.
Monoclonal
Flow Cytometry, Immunocytochemistry, Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-20 Receptor
IgG2a, kappa
RUO
Primary
Human, Murine