missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24165-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by IL-27 is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR).Specifications
IL-27 | |
Polyclonal | |
Unconjugated | |
IL27 | |
IL 27, IL 27 A, IL 27 subunit alpha, IL 27A, IL27, IL27 A, IL27A, IL27p28, IL30, interleukin 27, Interleukin 27 subunit alpha, p28 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246778 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NEV9 | |
IL27 | |
IL-27 Fusion Protein Ag21436 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |