Learn More
IL-28 (IFN lambda) Monoclonal Antibody (RN28), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1046.00 SEK - 2855.00 SEK
Specifications
Antigen | IL-28 (IFN lambda) |
---|---|
Clone | RN28 |
Concentration | 0.5 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15218057
|
Affymetrix eBioscience
14-7281-81 |
50 μg |
1046.00 SEK
50µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15228057
|
Affymetrix eBioscience
14-7281-85 |
500 μg |
2855.00 SEK
500µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This monoclonal RN28 antibody reacts with IL-28 (IFN lambda), a member of the IFN-lambda family which mediates the induction of anti-viral protection in a wide variety of cells. Applications Reported: The RN28 antibody has been reported for use in ELISA capture of mouse IL-28. Applications Tested: This RN28 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse IL-28 in combination with the biotin polyclonal (cat. 13-7282) antibody for detection. A suitable range of concentrations of this antibody for ELISA capture is 1.0-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 4000 pg/mL - 30 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
IL28 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).Specifications
IL-28 (IFN lambda) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Cytokine Zcyto20; cytokine Zcyto22; EG330496; IFL-1; IFN lambda2; Ifnl2; Ifnl3; IFN-lambda2; IFN-lambda-2; IFN-lambda-3; IFN-lambda-4; IL 28; IL 28 A; Il28; Il28a; IL-28A; Il28b; IL-28B; IL28C; IL-28C; ILN; INF-alpha; INF-lambda; interferon alpha; interferon lambda 2; interferon lambda 3; Interferon lambda2; interferon lambda-2; Interferon lambda-3; interferon, lambda 2; interferon, lambda 3; interferon, lambda 4; interferon-lambda 2; interferon-lambda2; Interleukin; interleukin 28; interleukin 28A; interleukin 28A (interferon, lambda 2); interleukin 28B; interleukin-28A; Interleukin-28B; interleukin-28C; ZCYTO20; ZCYTO22 | |
Ifnl2, Ifnl3 | |
Primary | |
4° C |
RN28 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q4VK74, Q8CGK6 | |
330496, 338374 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.