Läs mer
Invitrogen™ IL-4 Monoclonal Antibody (E10023)
Mouse Monoclonal Antibody
Brand: Invitrogen™ M3IL4I
Beskrivning
M3IL4I targets IL-4 in ELISA and Western blot applications and shows reactivity with Human samples. The M3IL4I immunogen is recombinant human IL-4 purified from Pichia pastoris. M3IL4I detects IL-4 which has a predicted molecular weight of approximately 15 kDa. The M3IL4I IL4 antibody (clone E10023) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M2IL4BI (biotinylated conjugate of clone E10022). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Antibody M3IL4I (clone E10022) and biotinylated antibody M2IL4BI (clone E10023) have successfully been used in combination with recombinant IL4 protein SIL4 in ELISA applications. Endotoxin levels are less than or equal to 10 μg/mL.
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Specifikationer
| IL-4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 10% trehalose and no preservative | |
| P05112 | |
| Il4 | |
| Recombinant human IL-4 purified from Pichia pastoris. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| E10023 | |
| Unconjugated | |
| Il4 | |
| B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
| Mouse | |
| Protein G | |
| RUO | |
| 3565 | |
| 4°C | |
| Lyophilized |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.