Learn More
IL-4 Monoclonal Antibody (8D4-8), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2555.00 SEK - 5850.00 SEK
Specifications
Antigen | IL-4 |
---|---|
Clone | 8D4-8 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16626384
|
Affymetrix eBioscience
48-7049-42 |
100 Tests |
5850.00 SEK
Pack of 100 |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16616384
|
Affymetrix eBioscience
48-7049-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 8D4-8 antibody reacts with human interleukin-4 (IL-4), a 15-19 kDa cytokine secreted by Th2 cells. Applications Reported: This 8D4-8 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 8D4-8 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to "e;Staining Intracellular Antigens for Flow Cytometry, Protocol A: Two step protocol for intracellular (cytoplasmic) proteins"e; located at <a href="e;https://www.thermofisher.com/flowprotocols"e;>www.thermofisher.com/flowprotocols</a>. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser
IL-4 Monoclonal antibody specifically detects IL-4 in Human samples. It is validated for Flow Cytometry.Specifications
IL-4 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
3565 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Il4 |
8D4-8 | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
P05112 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |