missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL15RA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16744-1-AP-20UL
Dettagli aggiuntivi : Weight : 0.00010kg
Descrizione
Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.Specifica
IL15RA | |
Polyclonal | |
Unconjugated | |
IL15RA | |
IL 15 receptor subunit alpha, IL 15R alpha, IL 15RA, IL15RA, interleukin 15 receptor, alpha, sIL 15 receptor subunit alpha, sIL 15R alpha, sIL 15RA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3601 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13261 | |
IL15RA | |
IL15RA Fusion Protein Ag10151 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |