missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F10 Mouse anti-Human, Clone: 5B2F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60319-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The cytokine IL-1 is responsible for initiating a variety of activities through the activation of transcription factors, NF-kappa B and AP-1, thereby promoting host response to injury or infection. IL-1F10 (Interleukin-1 family member 10), also known as FIL1T, IL1HY2 or FKSG75, is highly expressed in the skin, spleen and proliferating B-cells of the tonsil. IL-1F10 binds soluble IL-1 receptor type 1, and is one of nine Interleukin 1 families clustered on chromosome 2, where it is thought to participate in the regulation of adapted and innate immune responses. IL-1F10 and IL-1RN polymorphisms may play an important role in the susceptibility to developing rheumatoid arthritis.Specifications
IL1F10 | |
Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WWZ1 | |
IL1F10 | |
IL1F10 Fusion Protein Ag17250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
5B2F5 | |
Unconjugated | |
IL1F10 | |
FIL1 theta, FIL1T, FKSG75, IL 1 theta, IL 1F10, IL 1HY2, IL1 theta, IL1F10, IL1HY2, Interleukin 1 family member 10, Interleukin 1 HY2, Interleukin 1 theta, UNQ6119/PRO20041 | |
Mouse | |
Protein G | |
RUO | |
84639 | |
-20°C | |
Liquid |