missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F10 Mouse anti-Human, Clone: 5B2F5, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | IL1F10 |
---|---|
Clone | 5B2F5 |
Concentration | 2 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827415
|
Proteintech
60319-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817415
|
Proteintech
60319-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The cytokine IL-1 is responsible for initiating a variety of activities through the activation of transcription factors, NF-kappa B and AP-1, thereby promoting host response to injury or infection. IL-1F10 (Interleukin-1 family member 10), also known as FIL1T, IL1HY2 or FKSG75, is highly expressed in the skin, spleen and proliferating B-cells of the tonsil. IL-1F10 binds soluble IL-1 receptor type 1, and is one of nine Interleukin 1 families clustered on chromosome 2, where it is thought to participate in the regulation of adapted and innate immune responses. IL-1F10 and IL-1RN polymorphisms may play an important role in the susceptibility to developing rheumatoid arthritis.Specifications
IL1F10 | |
2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FIL1 theta, FIL1T, FKSG75, IL 1 theta, IL 1F10, IL 1HY2, IL1 theta, IL1F10, IL1HY2, Interleukin 1 family member 10, Interleukin 1 HY2, Interleukin 1 theta, UNQ6119/PRO20041 | |
IL1F10 | |
IgG1 | |
Protein G | |
Antibody |
5B2F5 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8WWZ1 | |
84639 | |
IL1F10 Fusion Protein Ag17250 | |
Primary | |
-20°C | |
IL1F10 |