missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-31 Recombinant Protein

Human IL-31 Recombinant Protein

Brand:  Gibco™ PHC0314

Additional Details : Weight : 0.02000kg

Product Code. 10619053

  • 5080.00 SEK / Each
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

Human IL-31 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL31 is primarily associated with activated T-cells and expressed by Th2 rather than Th1 cells. IL31 signals via IL31RA and the oncostatin M receptor. IL31 signaling has been shown to involve the Jak/STAT pathway, the PI3 kinase/AKT cascade, and MAP kinase pathway.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
Sequential Chromatography
IL-31
Human
Carrier-Free
10 μg
E. coli
Recombinant, Ligand
Interleukins
Gibco™
15.8 kDa
10 ug
141 aa
Store in freezer (-5 to -30°C).
< 0.1 ng/μg
Wet Ice
Cytokines & Receptors
Lyophilized
Inflammation, Immunology
98 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.